31454 Sigma-Aldrich

Sodium sulfite

puriss. p.a., Reag. Ph. Eur., anhydrous, 98-100.0% (iodometric)

DOWNLOAD MSDS (PDF)
  • CAS Number 7757-83-7

  • Linear Formula Na2SO3

  • Molecular Weight 126.04

  •  EC Number 231-821-4

  •  MDL number MFCD00003503

  •  PubChem Substance ID 24858963

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Puriss p.a., Q-S, Puriss p.a. More...
grade   Reag. Ph. Eur.
  puriss. p.a.
assay   98-100.0% (iodometric)
impurities   ≤0.001% heavy metals (as Pb)
  ≤0.02% thiosulfate (S2O3)
anion traces   chloride (Cl-): ≤50 mg/kg
cation traces   As: ≤0.2 mg/kg
  Ca: ≤50 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Se: ≤10 mg/kg
  Zn: ≤10 mg/kg

Price and Availability

Customers Also Viewed

Sodium metabisulfite

puriss. p.a., ACS reagent, reag. Ph. Eur., dry, 98-100.5%

Sodium sulfite

≥98%

Sodium sulfite

BioUltra, anhydrous, ≥98.0% (RT)

Sodium sulfite

puriss. p.a., ACS reagent, anhydrous, ≥98.0% (RT)

Sodium sulfite

purum p.a., anhydrous, ≥97.0% (RT)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?