31483 Sigma-Aldrich

Nickel(II) sulfate hexahydrate

puriss. p.a., ACS reagent, 99-102.0%

DOWNLOAD MSDS (PDF)
  • CAS Number 10101-97-0

  • Linear Formula NiSO4 · 6H2O

  • Molecular Weight 262.85

  •  EC Number 232-104-9

  •  MDL number MFCD00149813

  •  PubChem Substance ID 24858977

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, M-N, Puriss p.a. ACS, Puriss p.a. ACS More...
grade   ACS reagent
vapor density   2.07 (vs air)
grade   puriss. p.a.
assay   99-102.0%
impurities   ≤0.001% total nitrogen (N)
  ≤0.005% in water insoluble matters
pH   4-6 (20 °C, 5%)
anion traces   chloride (Cl-): ≤10 mg/kg
cation traces   Ca: ≤50 mg/kg
  Co: ≤20 mg/kg
  Cu: ≤20 mg/kg
  Fe: ≤10 mg/kg
  K: ≤50 mg/kg
  Mg: ≤50 mg/kg
  Mn: ≤5 mg/kg
  Na: ≤100 mg/kg
  Pb: ≤10 mg/kg
  Zn: ≤20 mg/kg
suitability   complies for identity (IR)

Description

General description

bluegreen to green crystals

Price and Availability

Customers Also Viewed

Nickel(II) sulfate

anhydrous, 99.99% trace metals basis

Nickel(II) sulfate heptahydrate

purum p.a., crystallized, ≥99.0% (KT)

Nickel(II) sulfate hexahydrate

purum p.a., crystallized, ≥99.0% (KT)

Nickel(II) sulfate hexahydrate

≥99.99% trace metals basis

Nickel(II) sulfate hexahydrate

ACS reagent, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?