EMAIL THIS PAGE TO A FRIEND
31547 Sigma-Aldrich

2,6-Diacetylpyridine

purum, ≥98.0% (GC)

  • CAS Number 1129-30-2

  • Empirical Formula (Hill Notation) C9H9NO2

  • Molecular Weight 163.17

  •  Beilstein Registry Number 118286

  •  EC Number 214-442-9

  •  MDL number MFCD00006304

  •  PubChem Substance ID 329753803

Purchase

Properties

Related Categories Building Blocks, C9, Chemical Synthesis, Heterocyclic Building Blocks, Pyridines More...
InChI Key   BEZVGIHGZPLGBL-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
mp   79-82 °C(lit.)
  79-82 °C

Description

Other Notes

This product has been replaced by D8801-ALDRICH | 2,6-Diacetylpyridine 99%

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?