EMAIL THIS PAGE TO A FRIEND
31683 Sigma-Aldrich

Diethyl ether

puriss., dried over molecular sieve Deperox Dehydrat (H2O ≤0.005%), ≥99.8% (GC)

Synonym: Ether, Ethyl ether

  • CAS Number 60-29-7

  • Linear Formula (CH3CH2)2O

  • Molecular Weight 74.12

  •  Beilstein Registry Number 1696894

  •  EC Number 200-467-2

  •  MDL number MFCD00011646

  •  PubChem Substance ID 57648600

Purchase

Properties

Related Categories Diethyl Ether, Solvent by Type, Solvents More...
vapor density   2.6 (vs air)
vapor pressure   2311 hPa ( 60 °C)
InChI Key   RTZKZFJDLAIYFH-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.8% (GC)
autoignition temp.   320 °F
quality   dried over molecular sieve Deperox Dehydrat (H2O ≤0.005%)
contains   ~0.0005% 2,6-di-tert-butyl-4-methylphenol as stabilizer
expl. lim.   36.5 %
impurities   ≤0.00003% peroxides (as H2O2)
color   colorless
refractive index   n20/D 1.3530(lit.)
  n20/D 1.353
bp   34.6 °C(lit.)
mp   −116 °C(lit.)
density   0.706 g/mL at 25 °C(lit.)

Description

Application

Diethyl ether may be employed as solvent for the oxidation of secondary alcohols into ketones. It may be employed as solvent in the preparation of triblock copolymer of PCL-PEG-PCL, acrylated PCL-PEG-PCL and the pentablock copolymer PAE-PCL-PEG-PCL-PAE [PAE = poly(β-amino ester), PEG = poly(ethylene glycol), PCL = poly(ε-caprolactone)].

General description

Diethyl ether (DEE) is widely employed as solvent for Grignard synthesis. Its efficacy as solvent for Formalin-ether (F-E) sedimentation technique has been studied. It has been reported as a hydroboration solvent. DEE is a promising compression-ignition fuel and its energy density is much higher than ethanol. It is also one of the components in a blend used in diesel fuel.

Price and Availability


Added X
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
May form explosive peroxides., Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN1155 - class 3 - PG 1 - Diethyl ether
WGK Germany 
1
RTECS 
KI5775000
Flash Point(F) 
-40 °F
Flash Point(C) 
-40 °C
Protocols & Articles

Related Content

Properties of Solvents Table

a. Gant, R., LC/GC 10(7), 515 (1992) 1. At 20 °C 2. At 20 °C +/- 5 °C relative to water at 4 °C 3. At 10mmHg pressure 4. At l=589.32nm
Keywords: Gas chromatography, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

recommended

Product #

Description

Add to Cart

Z683299 XPell indicating pellets for peroxides prevention, CE for organic solvents

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?