EMAIL THIS PAGE TO A FRIEND
31700 Sigma-Aldrich

Diethyl ether

purum, ≥99.0% (GC)

Synonym: Ether, Ethyl ether

  • CAS Number 60-29-7

  • Linear Formula (CH3CH2)2O

  • Molecular Weight 74.12

  •  Beilstein Registry Number 1696894

  •  EC Number 200-467-2

  •  MDL number MFCD00011646

  •  PubChem Substance ID 57648603

Purchase

Properties

Related Categories Diethyl Ether, Solvent by Type, Solvents More...
vapor density   2.6 (vs air)
InChI Key   RTZKZFJDLAIYFH-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
autoignition temp.   320 °F
contains   ~0.0005% 2,6-di-tert-butyl-4-methylphenol as stabilizer
expl. lim.   36.5 %
impurities   ≤0.2% water (Karl Fischer)
refractive index   n20/D 1.3530(lit.)
  n20/D 1.353
bp   34.6 °C(lit.)
mp   −116 °C(lit.)
density   0.706 g/mL at 25 °C(lit.)

Description

Other Notes

Sales restrictions may apply

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Application

Diethyl ether may be employed as a solvent for the oxidation of secondary alcohols into ketones. It may be employed as solvent in the preparation of triblock copolymer of PCL-PEG-PCL, acrylated PCL-PEG-PCL and the pentablock copolymer PAE-PCL-PEG-PCL-PAE [PAE = poly(β-amino ester), PEG = poly(ethylene glycol), PCL = poly(ε-caprolactone)].

General description

Diethyl ether (DEE) is widely employed as solvent for Grignard synthesis. Its efficacy as solvent for Formalin-ether (F-E) sedimentation technique has been studied. It has been reported as a hydroboration solvent. DEE is a promising compression-ignition fuel and its energy density is much higher than ethanol. It is also one of the components in a blend used in diesel fuel. It can also be used as alternative to diesel fuel.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
May form explosive peroxides., Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN1155 - class 3 - PG 1 - Diethyl ether
WGK Germany 
1
RTECS 
KI5775000
Flash Point(F) 
-40 °F
Flash Point(C) 
-40 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?