EMAIL THIS PAGE TO A FRIEND
31731 Sigma-Aldrich

Diethylamine

purum, ≥99.0% (GC)

  • CAS Number 109-89-7

  • Linear Formula (C2H5)2NH

  • Molecular Weight 73.14

  •  Beilstein Registry Number 605268

  •  EC Number 203-716-3

  •  MDL number MFCD00009032

Purchase

Properties

Related Categories Acids & Bases, Bases, Cell Biology, Chemical Synthesis, Hordeum vulgare (Barley),
vapor density   2.5 (vs air)
vapor pressure   14.14 psi ( 55 °C)
  180 mmHg ( 20 °C)
  3.51 psi ( 20 °C)
InChI Key   HPNMFZURTQLUMO-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
autoignition temp.   594 °F
expl. lim.   10.1 %
refractive index   n20/D 1.385(lit.)
  n20/D 1.386
bp   55 °C(lit.)
mp   −50 °C(lit.)
density   0.707 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Diethylamine was used for the modification of poly (glycidyl methacrylate) grafted resin and glycidyl methacrylate (GMA) - Ethylene glycol dimethacrylate (EGDMA) copolymer resin. It may be used to compose the basic solution for use in β-amyloid ELISA studies. It may be used in the rubella virus immunoglobulin G (IgG) avidity assay.

General description

Diethylamine is a secondary ethyl derivative of ammonia. It smells like ammonia. It is an important intermediate formed during the synthesis of resins and various pharmaceutical products.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1154 - class 3 - PG 2 - Diethylamine
WGK Germany 
1
RTECS 
HZ8750000
Flash Point(F) 
-9.4 °F
Flash Point(C) 
-23 °C
Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?