• USA Home
  • 31760 - 2-(Diethylamino)ethanol

EMAIL THIS PAGE TO A FRIEND
31760 Sigma-Aldrich

2-(Diethylamino)ethanol

purum, ≥99.0% (GC)

Synonym: N,N-Diethylethanolamine, DEAE, DEEA

  • CAS Number 100-37-8

  • Linear Formula (C2H5)2NCH2CH2OH

  • Molecular Weight 117.19

  •  Beilstein Registry Number 741863

  •  EC Number 202-845-2

  •  MDL number MFCD00002850

  •  PubChem Substance ID 24859090

Purchase

Properties

Related Categories Amino Alcohols, Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds More...
vapor density   4.04 (vs air)
vapor pressure   1 mmHg ( 20 °C)
InChI Key   BFSVOASYOCHEOV-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
expl. lim.   11.7 %
refractive index   n20/D 1.441(lit.)
  n20/D 1.442
bp   161 °C(lit.)
density   0.884 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2686 8(3) / PGII
WGK Germany 
1
RTECS 
KK5075000
Flash Point(F) 
122 °F
Flash Point(C) 
50 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?