• USA Home
  • 31810 - 2-Chloro-N,N-diethylethylamine hydrochloride

EMAIL THIS PAGE TO A FRIEND
31810 Sigma-Aldrich

2-Chloro-N,N-diethylethylamine hydrochloride

puriss., ≥99.0% (AT)

Synonym: 2-(Diethylamino)ethyl chloride hydrochloride, 2-Chlorotriethylamine hydrochloride

  • CAS Number 869-24-9

  • Linear Formula (C2H5)2NCH2CH2Cl · HCl

  • Molecular Weight 172.10

  •  Beilstein Registry Number 3591991

  •  EC Number 212-786-4

  •  MDL number MFCD00012519

  •  PubChem Substance ID 329753923

Purchase

Properties

InChI Key   RAGSWDIQBBZLLL-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (AT)
mp   208-210 °C(lit.)
  210-212 °C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2811 6.1/PG 2
WGK Germany 
3
RTECS 
YE1050000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?