319767 Sigma-Aldrich

Aluminum oxide

powder, -100 mesh, 99.9%

DOWNLOAD MSDS (PDF)

Synonym: Alumina

  • CAS Number 1344-28-1

  • Linear Formula Al2O3

  • Molecular Weight 101.96

  •  EC Number 215-691-6

  •  MDL number MFCD00003424

  •  PubChem Substance ID 24859187

Properties

assay   99.9%
form   powder
particle size   -100 mesh
mp   2040 °C(lit.)

Price and Availability

Customers Also Viewed

Aluminum oxide

Type CG-20

Aluminum oxide

puriss., ≥98% Al2O3 basis

Aluminum oxide

nanopowder, <50 nm particle size (TEM)

Aluminum oxide

99.998% trace metals basis

Aluminum oxide

powder, 99.99% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?