319953 Sigma-Aldrich

Benzene

ACS reagent, ≥99.0%

DOWNLOAD MSDS (PDF)

Properties

Related Categories ACS Grade, ACS Grade Solvents, ACS and Reagent Grade Solvents, Amber Glass Bottles, Benzene and Benzene Solutions,
grade   ACS reagent
vapor density   2.77 (vs air)
vapor pressure   166 mmHg ( 37.7 °C)
  74.6 mmHg ( 20 °C)
assay   ≥99.0%
autoignition temp.   1043 °F
expl. lim.   8 %
impurities   H2SO4, passes test(darkened)
  thiophene, passes test(limit ∼1ppm)
  ≤0.005% S compounds
  ≤0.05% water (Karl Fischer)
evapn. residue   ≤0.001%
color   APHA: ≤10
refractive index   n20/D 1.501(lit.)
bp   80 °C(lit.)
mp   5.5 °C(lit.)
density   0.874 g/mL at 25 °C(lit.)

Description

Packaging

1, 2, 2.5, 4, 4×4 L in glass bottle

20 L in steel drum

500 mL in glass bottle

Price and Availability

Customers Also Viewed

Benzene
270709Benzene

CHROMASOLV® Plus, for HPLC, ≥99.9%

Benzene
12540Benzene

analytical standard

Benzene
401765Benzene

anhydrous, 99.8%

Benzene
32212Benzene

puriss. p.a., Reag. Ph. Eur., ≥99.7% (GC)

Benzene
676985Benzene

ACS reagent, ≥99.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?