319988 Sigma-Aldrich

Chloroform

contains ethanol as stabilizer, ACS reagent, ≥99.8%

DOWNLOAD MSDS (PDF)

Synonym: Methylidyne trichloride, Trichloromethane

Properties

Related Categories ACS Grade, ACS Grade Solvents, ACS and Reagent Grade Solvents, Amber Glass Bottles, Carbon Steel Cans with NPT Threads,
grade   ACS reagent
vapor density   4.1 (vs air)
vapor pressure   160 mmHg ( 20 °C)
assay   ≥99.8%
contains   ethanol as stabilizer
impurities   Acetone and aldehyde, passes test
  Acid and chloride, passes test
  Free chlorine (Cl), passes test
  H2SO4, passes test
evapn. residue   ≤0.001%
color   APHA: ≤10
refractive index   n20/D 1.445(lit.)
bp   60.5-61.5 °C(lit.)
mp   −63 °C(lit.)
density   1.48 g/mL at 25 °C
  1.492 g/mL at 25 °C(lit.)
cation traces   Pb: ≤0.05 ppm

Description

Frequently Asked Questions

Live Chat and Frequently Asked Questions are available for this Product.

Packaging

1, 6×1, 2, 4×2, 2.5, 4×2.5, 4×4 L in glass bottle

20, 200 L in steel drum

500, 6×500 mL in glass bottle

Price and Availability

Customers Also Viewed

Chloroform

contains 100-200 ppm amylenes as stabilizer, ≥99.5%

Chloroform

anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer

Chloroform

ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer

Chloroform

contains amylenes as stabilizer, ACS reagent, ≥99.8%

Chloroform

ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?