CAS Number 7732-18-5
Empirical Formula (Hill Notation) H2O
Molecular Weight 18.02
Beilstein Registry Number 2050024
EC Number 231-791-2
MDL number MFCD00011332
PubChem Substance ID 24859243
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade Solvents, ACS and Reagent Grade Solvents, Biological Buffers, Buffers A to Z, Plastic Bottles, |
| grade | ACS reagent |
| impurities | MnO4- reducers, passes test |
| ≤0.01% Silicate (as SiO2) | |
| ≤0.01 ppm silicate | |
| refractive index |
n |
| conductivity | ≤2 μmho per cm at 25 °C |
| bp | 100 °C(lit.) |
2, 4×2, 2.5, 4×4 L in poly bottle
200 L in poly drum
500 mL in poly bottle
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?