CAS Number 7647-01-0
Empirical Formula (Hill Notation) HCl
Molecular Weight 36.46
Beilstein Registry Number 1098214
EC Number 231-595-7
MDL number MFCD00011324
PubChem Substance ID 24859260
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade, ACS Grade Acids & Bases, Acids & Bases, Analytical Reagents, Analytical Reagents for General Use, |
| grade | ACS reagent |
| vapor density | 1.3 (vs air) |
| vapor pressure | 3.23 psi ( 21.1 °C) |
| 7.93 psi ( 37.7 °C) | |
| assay | 36.5-38.0% (ACS specification) |
| 37% | |
| impurities | ≤1 ppm free chlorine |
Live Chat and Frequently Asked Questions
are available for this Product.
2.5, 6×2.5 L in PVC coated
500, 6×500 mL in PVC coated
Eliminate repetitive Molar calculations and potential errors with these conveniently designed graduated cylinders. They featureCustomers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?