EMAIL THIS PAGE TO A FRIEND
320501 Sigma-Aldrich

Sulfuric acid

ACS reagent, 95.0-98.0%

Purchase

Properties

Related Categories ACS Grade Acids & Bases, Acids, Acids & Bases, Adsorbents, Filter Aids and Drying Agents, Analytical Reagents,
grade   ACS reagent
vapor density   <0.3 (25 °C, vs air)
vapor pressure   1 mmHg ( 146 °C)
description   Nominally 95-98% H2SO4
assay   95.0-98.0%
appearance   Free from suspended or insoluble matter
ign. residue   ≤5 ppm
color   APHA: ≤10
bp   ~290 °C(lit.)
density   1.840 g/mL at 25 °C(lit.)
anion traces   MnO4- reducers: ≤2 ppm
  chloride (Cl-): ≤0.2 ppm
  nitrate (NO3-): ≤0.5 ppm
cation traces   As: ≤0.01 ppm
  Fe: ≤0.2 ppm
  Hg: ≤5 ppb
  NH4+: ≤2 ppm
  heavy metals (as Pb): ≤1 ppm

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Packaging

2.5, 6×2.5 L in PVC-coated bottle

500, 6×500 mL in PVC-coated bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1830 8 / PGII
WGK Germany 
1
RTECS 
WS5600000
Protocols & Articles

Articles

Determination of Water Content in Sulfuric acid Using Karl Fischer Titration

Strong acids shift the pH of the KF reagent and must be neutralised. This can be carried out directly in thetitration cell providing a sufficient amount of buffer is added, e.g. HYDRANAL-Buffer Acid,...
Keywords: Esterifications, Titrations

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?