EMAIL THIS PAGE TO A FRIEND
32080 Sigma-Aldrich

Diethyl carbonate

purum, ≥98.0% (GC)

  • CAS Number 105-58-8

  • Linear Formula (C2H5O)2CO

  • Molecular Weight 118.13

  •  Beilstein Registry Number 956591

  •  EC Number 203-311-1

  •  MDL number MFCD00009107

  •  PubChem Substance ID 24859279

Purchase

Properties

Related Categories Chemical Synthesis, Other Protecting and Derivatizing Reagents, Protecting and Derivatizing Reagents, Protection and Derivatization, Synthetic Reagents More...
vapor density   4.1 (vs air)
vapor pressure   10 mmHg ( 23.8 °C)
  59 mmHg ( 37.8 °C)
InChI Key   OIFBSDVPJOWBCH-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.384(lit.)
  n20/D 1.384
bp   126-128 °C(lit.)
mp   −43 °C(lit.)
density   0.975 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2366 3 / PGIII
WGK Germany 
1
RTECS 
FF9800000
Flash Point(F) 
77 °F
Flash Point(C) 
25 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?