• USA Home
  • 32085 - Diethyl chlorophosphate

EMAIL THIS PAGE TO A FRIEND
32085 Sigma-Aldrich

Diethyl chlorophosphate

purum, >97.0% (T)

Synonym: Diethyl phosphorochloridate

  • CAS Number 814-49-3

  • Linear Formula (C2H5O)2P(O)Cl

  • Molecular Weight 172.55

  •  Beilstein Registry Number 471433

  •  EC Number 212-396-4

  •  MDL number MFCD00009075

  •  PubChem Substance ID 24859280

Purchase

Properties

vapor density   5.94 (vs air)
vapor pressure   0.1 mmHg ( 25 °C)
InChI Key   LGTLXDJOAJDFLR-UHFFFAOYSA-N
grade   purum
assay   >97.0% (T)
refractive index   n20/D 1.416(lit.)
bp   60 °C/2 mmHg(lit.)
density   1.194 g/mL at 25 °C(lit.)

Description

Other Notes

This product has been replaced by D91632-ALDRICH | Diethyl chlorophosphate 97%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3278 6.1 / PGI
WGK Germany 
3
Flash Point(F) 
199.4 °F
Flash Point(C) 
93 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?