• USA Home
  • 32207 - Pentanol mixture of isomers

EMAIL THIS PAGE TO A FRIEND
32207 Sigma-Aldrich

Pentanol mixture of isomers

for determination of fat in milk according to Gerber

Synonym: Amyl alcohol mixture of isomers

  • CAS Number 30899-19-5

  • Empirical Formula (Hill Notation) C5H12O

  • Molecular Weight 88.15

  •  Beilstein Registry Number 1730975

  •  EC Number 250-378-8

  •  MDL number MFCD00002977

Purchase

Properties

grade   for determination of fat in milk according to Gerber
InChI Key   AMQJEAYHLZJPGS-UHFFFAOYSA-N
impurities   ≤0.003% non-volatile matter
  ≤0.3% water (Karl Fischer)
  ≥99% total isomers (GC)
refractive index   n20/D 1.4065-1.4085
density   0.808-0.814 g/mL at 20 °C
suitability   passes test for reaction against H2SO4
  passes test for suitability for fat test acc Gerber

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN1105 - class 3 - PG 3 - Pentanols
WGK Germany 
1
Flash Point(F) 
114.8 °F
Flash Point(C) 
46 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?