32249 Sigma-Aldrich

Toluene

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)

DOWNLOAD MSDS (PDF)
  • CAS Number 108-88-3

  • Linear Formula C6H5CH3

  • Molecular Weight 92.14

  •  Beilstein Registry Number 635760

  •  EC Number 203-625-9

  •  MDL number MFCD00008512

  •  PubChem Substance ID 24859319

Properties

Related Categories ACS Grade, Amber Glass Bottles, Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography,
grade   ACS reagent, reag. ISO, reag. Ph. Eur.
vapor density   3.2 (vs air)
vapor pressure   22 mmHg ( 20 °C)
  26 mmHg ( 25 °C)
grade   puriss. p.a.
assay   ≥99.7% (GC)
autoignition temp.   997 °F
expl. lim.   7 %
impurities   ≤0.0001% thiophene
  ≤0.001% free acid (as HCl)
  ≤0.001% free alkali (as NaOH)
  ≤0.001% non-volatile matter
  ≤0.003% S-compounds (as S)
  ≤0.02% water (Karl Fischer)
color   APHA: ≤10
refractive index   n/D 1.496(lit.)
  n20/D 1.496-1.498
bp   110-111 °C(lit.)
mp   -93 °C(lit.)
density   0.865 g/mL at 25 °C(lit.)
cation traces   Al: ≤0.5 ppm
  Ba: ≤0.1 ppm
  Bi: ≤0.1 ppm
  Ca: ≤0.5 ppm
  Cd: ≤0.05 ppm
  Co: ≤0.02 ppm
  Cr: ≤0.02 ppm
  Cu: ≤0.02 ppm
  Fe: ≤0.1 ppm
  K: ≤0.5 ppm
  Li: ≤0.1 ppm
  Mg: ≤0.1 ppm
  Mn: ≤0.02 ppm
  Mo: ≤0.1 ppm
  Na: ≤0.5 ppm
  Ni: ≤0.02 ppm
  Pb: ≤0.1 ppm
  Sr: ≤0.1 ppm
  Zn: ≤0.1 ppm

Description

Other Notes

Pertains only in Germany: unversteuert

Price and Availability

Customers Also Viewed

Acetone
32201Acetone

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)

Hexane
32293Hexane

puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)

Methanol

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)

Toluene
244511Toluene

anhydrous, 99.8%

Toluene
34938Toluene

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., dried, ≥99.7% (GC), H2O <0.001%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?