• USA Home
  • 32270 - Diethylene glycol monohexyl ether

EMAIL THIS PAGE TO A FRIEND
32270 Sigma-Aldrich

Diethylene glycol monohexyl ether

purum, ≥95.0% (GC)

Synonym: 2-(2-Hexyloxyethoxy)ethanol, C6E2, Hexyldiglycol

  • CAS Number 112-59-4

  • Linear Formula CH3(CH2)5OCH2CH2OCH2CH2OH

  • Molecular Weight 190.28

  •  Beilstein Registry Number 1743959

  •  EC Number 203-988-3

  •  MDL number MFCD00010703

  •  PubChem Substance ID 329754115

Purchase

Properties

Related Categories Biochemicals and Reagents, Detergents, Detergents A to M, Non-Ionic Detergents
description   non-ionic
InChI Key   GZMAAYIALGURDQ-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
refractive index   n20/D 1.438
bp   261-265 °C(lit.)
density   0.932 g/mL at 20 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
WGK Germany 
1
RTECS 
KL2625000
Flash Point(F) 
284 °F
Flash Point(C) 
140 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

449393 Di(ethylene glycol) hexyl ether, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?