• USA Home
  • 323195 - 2-Pyridinecarboxaldehyde 4-nitrophenylhydrazone

EMAIL THIS PAGE TO A FRIEND
323195 Sigma-Aldrich

2-Pyridinecarboxaldehyde 4-nitrophenylhydrazone

97%

  • CAS Number 70421-66-8

  • Empirical Formula (Hill Notation) C12H10N4O2

  • Molecular Weight 242.23

  •  MDL number MFCD00012100

  •  PubChem Substance ID 24859226

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, P, Stains & Dyes, A to Z, Stains and Dyes More...
InChI Key   JXFHYUCJPGWCFI-NTEUORMPSA-N
assay   97%
mp   256-259 °C(lit.)

Description

Application

pH indicator and Pd reagent.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?