• USA Home
  • 32445 - Diethylphenylphosphine

EMAIL THIS PAGE TO A FRIEND
32445 Sigma-Aldrich

Diethylphenylphosphine

purum, ≥97.0% (GC)

Synonym: NSC 158475

  • CAS Number 1605-53-4

  • Linear Formula (C2H5)2PC6H5

  • Molecular Weight 166.20

  •  Beilstein Registry Number 742484

  •  EC Number 216-516-6

  •  MDL number MFCD00015172

Purchase

Properties

InChI Key   LVTCZSBUROAWTE-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.546(lit.)
bp   120-121 °C/29 mmHg(lit.)
density   0.954 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Catalyst for:
• Selective cross-dimerization
• Diastereoselective condensations
• Carboxyl migration reactions
• Selective hydrogenation
• Asymmetric induction by chiral diphosphines in ring contraction

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993 / PGIII
WGK Germany 
3
Flash Point(F) 
183.2 °F
Flash Point(C) 
84 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

381241 Diethylphenylphosphine, 96%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?