EMAIL THIS PAGE TO A FRIEND
32530 Sigma-Aldrich

Diethyl sulfide

puriss., ≥99.0% (GC)

Synonym: 1,1′-Thiobisethane, Ethyl sulfide, NSC 75157

Purchase

Properties

Related Categories Building Blocks, Cell Biology, Chemical Synthesis, Nutrition Research, Organic Building Blocks,
vapor pressure   105 mmHg ( 37.7 °C)
InChI Key   LJSQFQKUNVCTIA-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (GC)
refractive index   n20/D 1.442(lit.)
  n20/D 1.442
bp   90-92 °C(lit.)
mp   −100 °C(lit.)
density   0.837 g/mL at 25 °C(lit.)

Description

Application

Diethyl sulfide was employed as ancillary ligand in the preparation of brightly luminescent Pt(II) pincer complexes.

General description

Diethyl sulfide is a simulant for chemical agent mustard gas. Photooxidation of gaseous diethyl sulfide over TiO2 in a flow reactor was investigated.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2375 3 / PGII
WGK Germany 
3
RTECS 
LC7200000
Flash Point(F) 
15.8 °F
Flash Point(C) 
-9 °C
Protocols & Articles

Articles

GC Analysis of a 12-Component Sulfur Compound Mix on SLB®-IL59

From our library of Articles, Sigma-Aldrich presents GC Analysis of a 12-Component Sulfur Compound Mix on SLB®-IL59
Keywords: Chromatography, Gas chromatography, Mass selective detector, Size-exclusion chromatography

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

107247 Diethyl sulfide, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?