• USA Home
  • 32748 - 3,3′-Diaminobenzidine tetrahydrochloride

EMAIL THIS PAGE TO A FRIEND
32748 Sigma-Aldrich

3,3′-Diaminobenzidine tetrahydrochloride

tablet, 10 mg substrate per tablet

Synonym: 3,3′,4,4′-Biphenyltetramine tetrahydrochloride, 3,3′,4,4′-Tetraaminobiphenyl tetrahydrochloride, DAB

  • Linear Formula (NH2)2C6H3C6H3(NH2)2 · 4HCl

  • Molecular Weight 360.11

  •  Beilstein Registry Number 1212988

  •  EC Number 231-018-9

  •  MDL number MFCD00012969

  •  PubChem Substance ID 329754313

Purchase

Properties

Related Categories Biochemicals and Reagents, Fluorescent Enzyme Substrates, Fluorescent Enzyme Substrates for Electrophoresis, Fluorescent Enzyme Substrates for Microscopy, Fluorescent Probes, Labels, Particles and Stains More...
InChI Key   KJDSORYAHBAGPP-UHFFFAOYSA-N
form   tablet
packaging   set of 50 tablets
shipped in   wet ice
storage temp.   −20°C

Description

Application

Reagent for spectrophotometric determination of selenium.

Specificity

Peroxidase substrate suitable for use in immunoblotting and immunohistological staining procedures. This substrate produces an insoluble end product that is brown in color and can be observed visually.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DV8753000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?