EMAIL THIS PAGE TO A FRIEND
32791 Sigma-Aldrich

1,4-Diaminobutane

purum, ≥98.0% (GC)

Synonym: 1,4-Butanediamine, Putrescine, Tetramethylenediamine

  • CAS Number 110-60-1

  • Linear Formula NH2(CH2)4NH2

  • Molecular Weight 88.15

  •  Beilstein Registry Number 605282

  •  EC Number 203-782-3

  •  MDL number MFCD00008235

  •  PubChem Substance ID 24859703

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks, Polyamines More...
InChI Key   KIDHWZJUCRJVML-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
expl. lim.   9.08 %
impurities   ≤0.5% water
refractive index   n20/D 1.457(lit.)
bp   158-160 °C(lit.)
mp   25-28 °C(lit.)
density   0.877 g/mL at 25 °C(lit.)
Gene Information   rat ... Odc1(24609)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2925 8(4.1) / PGII
WGK Germany 
3
RTECS 
EJ6800000
Flash Point(F) 
123.8 °F
Flash Point(C) 
51 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?