EMAIL THIS PAGE TO A FRIEND
32908 Sigma-Aldrich

Cresol Red solution

acid-base indicator

  • CAS Number 1733-12-6

  • Empirical Formula (Hill Notation) C21H18O5S

  • Molecular Weight 382.43

  •  Beilstein Registry Number 343399

  •  MDL number MFCD00005878

  •  PubChem Substance ID 329754382

Purchase

Properties

grade   acid-base indicator
InChI Key   OBRMNDMBJQTZHV-UHFFFAOYSA-N
suitability   corresponds for change range
  corresponds for composition
  corresponds for sensitivity

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN1170 - class 3 - PG 3 - Ethanol, solution
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?