EMAIL THIS PAGE TO A FRIEND
32914 Sigma-Aldrich

T-2-Tetraol solution

50 μg/mL in acetonitrile, analytical standard

Synonym: 3α,4β,8α,15-Tetrahydroxy-12,13-epoxytrichothec-9-ene

  • CAS Number 34114-99-3

  • Empirical Formula (Hill Notation) C15H22O6

  • Molecular Weight 298.33

  •  EC Number 200-835-2

Purchase

Properties

Related Categories All alphabetically sorted, Analytical Standards, Analytical/Chromatography, Biotoxin and Mycotoxin Standards, Food and Cosmetic Component Standards,
grade   analytical standard
shelf life   limited shelf life, expiry date on the label
availability   not available in USA
concentration   50 μg/mL in acetonitrile
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   −20°C

Description

General description

Certan Vial

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1648 3 / PGII
WGK Germany 
2
Flash Point(F) 
42.8 °F
Flash Point(C) 
6 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?