• USA Home
  • 32975 - 2,3,5,6-Tetramethyl-p-phenylenediamine

EMAIL THIS PAGE TO A FRIEND
32975 Sigma-Aldrich

2,3,5,6-Tetramethyl-p-phenylenediamine

purum, ≥99.0% (NT)

Synonym: 3,6-Diaminodurene

  • CAS Number 3102-87-2

  • Linear Formula (CH3)4C6(NH2)2

  • Molecular Weight 164.25

  •  Beilstein Registry Number 2208743

  •  EC Number 221-457-4

  •  MDL number MFCD00007905

  •  PubChem Substance ID 24859811

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks, Polyamines More...
InChI Key   WCZNKVPCIFMXEQ-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (NT)
mp   150-154 °C
  150-155 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

523755 2,3,5,6-Tetramethyl-p-phenylenediamine, electronic grade, 99% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?