EMAIL THIS PAGE TO A FRIEND
33045 Sigma-Aldrich

Benzoic acid

for calorimetrical determination (approx. 26460 J/g)

  • CAS Number 65-85-0

  • Linear Formula C6H5COOH

  • Molecular Weight 122.12

  •  Beilstein Registry Number 636131

  •  EC Number 200-618-2

  •  MDL number MFCD00002398

  •  PubChem Substance ID 329754444

  •  eCl@ss 39023903

Purchase

Properties

Related Categories Acids, Acids & Bases, Benzoic Acid and Benzoic Acid Solutions, Chemical Synthesis, Essential Chemicals,
vapor density   4.21 (vs air)
vapor pressure   10 mmHg ( 132 °C)
InChI Key   WPYMKLBDIGXBTP-UHFFFAOYSA-N
form   crystalline
autoignition temp.   1061 °F
quality   for calorimetrical determination (approx. 26460 J/g)
bp   249 °C(lit.)
mp   121-125 °C(lit.)
solubility   water: soluble (2.9 g/l at 25 °C)

Description

Application

Benzoic acid may be employed as a standard in the quantitative and calorimetric studies. It may be employed as an intermediate in the synthesis of following products:
• paints
• pigments
• varnish
• wetting agents
• aroma compounds
• benzoyl chloride
• benzotrichloride

General description

Benzoic acid is an organic aromatic monocarboxylic acid. It occurs in the form of colorless leaflets or needles. It reacts with hydrogenating reagents to afford hexahydrobenzoic acid. On decomposition (by heating) in the presence of lime or alkali, it affords benzene and carbon dioxide. It can be synthesized by the cobalt or manganese catalyzed atmospheric oxidation of toluene.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Target organs 
Lungs
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
1
RTECS 
DG0875000
Flash Point(F) 
249.8 °F
Flash Point(C) 
121 °C
Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?