EMAIL THIS PAGE TO A FRIEND
33108 Sigma-Aldrich

Chromeazurol S

indicator for metal titration

Synonym: Chrome Azurol S, Mordant Blue 29

  • CAS Number 1667-99-8

  • Empirical Formula (Hill Notation) C23H13Cl2Na3O9S

  • Molecular Weight 605.28

  •  Beilstein Registry Number 4110655

  •  Colour Index Number 43825

  •  EC Number 216-787-0

  •  MDL number MFCD00001615

  •  PubChem Substance ID 24859932

Purchase

Properties

grade   indicator for metal titration
InChI Key   FUIZKNBTOOKONL-NDOZRWCBSA-K

Description

General description

Visit our Titration Center to learn more.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?