• USA Home
  • 33135 - 3,3′-Dimethylnaphthidine

EMAIL THIS PAGE TO A FRIEND
33135 Sigma-Aldrich

3,3′-Dimethylnaphthidine

indicator for metal titration

Synonym: 4,4′-Diamino-3,3′-dimethyl-1,1′-binaphthyl

  • CAS Number 13138-48-2

  • Empirical Formula (Hill Notation) C22H20N2

  • Molecular Weight 312.41

  •  Beilstein Registry Number 2152306

  •  EC Number 236-069-0

  •  MDL number MFCD00042706

  •  PubChem Substance ID 24859955

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Indicators, Metal Titration Indicators, Titration More...
grade   indicator for metal titration
InChI Key   FTEGMIZQLGXBNA-UHFFFAOYSA-N

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?