EMAIL THIS PAGE TO A FRIEND
33421 Sigma-Aldrich

m-Xylylenediamine

purum, ≥98.0% (NT)

Synonym: 1,3-Bis(aminomethyl)benzene, α,α′-Diamino-m-xylene

  • CAS Number 1477-55-0

  • Linear Formula C6H4(CH2NH2)2

  • Molecular Weight 136.19

  •  Beilstein Registry Number 1099911

  •  EC Number 216-032-5

  •  MDL number MFCD00008119

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks, Polyamines More...
vapor pressure   15 mmHg ( 145 °C)
InChI Key   FDLQZKYLHJJBHD-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (NT)
refractive index   n20/D 1.571(lit.)
  n20/D 1.572
bp   265 °C/745 mmHg(lit.)
density   1.032 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Corrosive to the respiratory tract.
RIDADR 
UN 2735PSN2 8 / PGII
WGK Germany 
2
RTECS 
PF8970000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles

Related Content

Carcinogen Selector - Digestive System - Liver

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Digestive System - Liver
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System - Liver Endocrine

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Endocrine System - Liver Endocrine
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Endocrine System
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Reproductive System - Lung

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Reproductive System - Lung
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Respiratory System

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Respiratory System
Keywords: Cancer, Carcinogens, Respiratory

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

X1202 m-Xylylenediamine, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?