MDL number MFCD00131613
Popular Documents: Specification Sheet (PDF)
| form | powder |
100, 500 g in poly bottle
Customers Also Viewed
BCR® certified Reference Material
BCR® certified Reference Material
powder, Catalyst support, ammonium Y zeolite
granular, +20 mesh
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?