EMAIL THIS PAGE TO A FRIEND
33532 Sigma-Aldrich

Dibenzenesulfonimide

purum, ≥97.0% (HPLC)

Synonym: Bis(phenylsulfonyl)amine, Dibenzenesulfimide

  • CAS Number 2618-96-4

  • Linear Formula (C6H5SO2)2NH

  • Molecular Weight 297.35

  •  Beilstein Registry Number 2145601

  •  EC Number 220-051-4

  •  MDL number MFCD00025019

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonimides/Sulfinimides, Sulfur Compounds More...
InChI Key   OVQABVAKPIYHIG-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (HPLC)
mp   150-155 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?