• USA Home
  • 338818 - Ammonium hydroxide solution

EMAIL THIS PAGE TO A FRIEND
338818 Sigma-Aldrich

Ammonium hydroxide solution

28% NH3 in H2O, ≥99.99% trace metals basis

Synonym: Ammonia aqueous, Ammonia water

Purchase

Properties

Related Categories Acids & Bases, Bases, Chemical Synthesis, Inorganic Bases, Synthetic Reagents More...
Quality Level   PREMIUM
assay   ≥99.99% trace metals basis
expl. lim.   27 %
concentration   28% NH3 in H2O
density   0.9 g/mL at 25 °C(lit.)
  0.9 g/mL at 25 °C

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Packaging

1 L in glass bottle

5, 100 mL in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2672 8 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Nanosized silver (II) pyridoxine complex to cause greater inflammatory response and less cytotoxicity to RAW264.7 macrophage cells. Paul A, Ju H, Rangasamy S, et al. Nanoscale Res. Lett. 10, 140, (2015)

Loading...


Comparative evaluation of seven different sample treatment approaches for large-scale multiclass sport drug testing in urine by liquid chromatography-mass spectrometry. Domínguez-Romero JC, García-Reyes JF, Molina-Díaz A, et al. J. Chromatogr. A. 1361, 34-42, (2014)

Loading...


Pharmacokinetic and pharmacodynamic analysis of inosine monophosphate dehydrogenase activity in hematopoietic cell transplantation recipients treated with mycophenolate mofetil. Li H, Mager DE, Sandmaier BM, et al. Biol. Blood Marrow Transplant. 20(8), 1121-9, (2014)

Loading...


High-performance electrocatalysis using metallic cobalt pyrite (CoS?) micro- and nanostructures. Faber MS, Dziedzic R, Lukowski MA, et al. J. Am. Chem. Soc. 136(28), 10053-61, (2014)

Loading...


Ultra high performance supercritical fluid chromatography coupled with tandem mass spectrometry for screening of doping agents. I: Investigation of mobile phase and MS conditions. Nováková L, Grand-Guillaume Perrenoud A, Nicoli R, et al. Anal. Chim. Acta 853, 637-46, (2014)

Loading...


Large-scale analysis of lysine SUMOylation by SUMO remnant immunoaffinity profiling. Lamoliatte F, Caron D, Durette C, et al. Nat. Commun. 5, 5409, (2014)

Loading...


Tracking targeted bimodal nanovaccines: immune responses and routing in cells, tissue, and whole organism. Cruz LJ, Tacken PJ, Zeelenberg IS, et al. Mol. Pharm. 11(12), 4299-313, (2014)

Loading...


Hydrophilic interaction chromatography versus reversed phase liquid chromatography coupled to mass spectrometry: effect of electrospray ionization source geometry on sensitivity. Periat A, Kohler I, Bugey A, et al. J. Chromatogr. A. 1356, 211-20, (2014)

Loading...


Quantification of cocaine and metabolites in exhaled breath by liquid chromatography-high-resolution mass spectrometry following controlled administration of intravenous cocaine. Ellefsen KN, Concheiro M, Beck O, et al. Anal. Bioanal. Chem 406(25), 6213-23, (2014)

Loading...


Comparison of liquid chromatography and supercritical fluid chromatography coupled to compact single quadrupole mass spectrometer for targeted in vitro metabolism assay. Spaggiari D, Mehl F, Desfontaine V, et al. J. Chromatogr. A. 1371, 244-56, (2014)

Loading...


Synthesis and in vitro localization study of curcumin-loaded SPIONs in a micro capillary for simulating a targeted drug delivery system. Anwar M, Asfer M, Prajapati AP, et al. Int. J. Pharm. 468(1-2), 158-64, (2014)

Loading...


Monitoring the effects of dexamethasone treatment by MRI using in vivo iron oxide nanoparticle-labeled macrophages. Gramoun A, Crowe LA, Maurizi L, et al. Arthritis. Res. Ther. 16(3), R131, (2014)

Loading...


Untargeted profiling of urinary steroid metabolites after testosterone ingestion: opening new perspectives for antidoping testing. Boccard J, Badoud F, Jan N, et al. Bioanalysis 6(19), 2523-36, (2014)

Loading...


Simultaneous analysis for water- and fat-soluble vitamins by a novel single chromatography technique unifying supercritical fluid chromatography and liquid chromatography. Taguchi K, Fukusaki E, Bamba T, et al. J. Chromatogr. A. 1362, 270-7, (2014)

Loading...


Simultaneous quantification of purine and pyrimidine bases, nucleosides and their degradation products in bovine blood plasma by high performance liquid chromatography tandem mass spectrometry. Stentoft C, Vestergaard M, Løvendahl P, et al. J. Chromatogr. A. 1356, 197-210, (2014)

Loading...


Determination of bisphenol A, triclosan and their metabolites in human urine using isotope-dilution liquid chromatography-tandem mass spectrometry. Provencher G, Bérubé R, Dumas P, et al. J. Chromatogr. A. 1348, 97-104, (2014)

Loading...


A high-throughput method for determination of metabolites of organophosphate flame retardants in urine by ultra performance liquid chromatography-high resolution mass spectrometry. Cequier E, Marcé RM, Becher G, et al. Anal. Chim. Acta 845, 98-104, (2014)

Loading...


Decreased plasma iron in Alzheimer's disease is due to transferrin desaturation. Hare DJ, Doecke JD, Faux NG, et al. ACS Chem. Neurosci. 6(3), 398-402, (2015)

Loading...


Au nanomatryoshkas as efficient near-infrared photothermal transducers for cancer treatment: benchmarking against nanoshells. Ayala-Orozco C, Urban C, Knight MW, et al. ACS Nano 8(6), 6372-81, (2014)

Loading...


Engineering Poly(ethylene glycol) Particles for Improved Biodistribution. Cui J, De Rose R, Alt K, et al. ACS Nano 9(2), 1571-80, (2015)

Loading...


Extracellular matrices decellularized from embryonic stem cells maintained their structure and signaling specificity. Sart S, Ma T, Li Y, et al. Tissue Eng. Part A 20(1-2), 54-66, (2014)

Loading...


The proteomic landscape of the suprachiasmatic nucleus clock reveals large-scale coordination of key biological processes. Chiang CK, Mehta N, Patel A, et al. PLoS Genet. 10(10), e1004695, (2014)

Loading...


Detection of melamine in milk using molecularly imprinted polymers-surface enhanced Raman spectroscopy. Hu Y, Feng S, Gao F, et al. Food Chem. 176, 123-9, (2015)

Loading...


Photocatalytic colour switching of redox dyes for ink-free light-printable rewritable paper. Wang W, Xie N, He L, et al. Nat. Commun. 5, 5459, (2014)

Loading...


Discovery of dual leucine zipper kinase (DLK, MAP3K12) inhibitors with activity in neurodegeneration models. Patel S, Cohen F, Dean BJ, et al. J. Med. Chem. 58(1), 401-18, (2015)

Loading...


Chemical hazards present in liquids and vapors of electronic cigarettes. Hutzler C, Paschke M, Kruschinski S, et al. Arch. Toxicol. 88(7), 1295-308, (2014)

Loading...


Bone marrow stromal cells enhance the osteogenic properties of hydroxyapatite scaffolds by modulating the foreign body reaction. Tour G, Wendel M, Tcacencu I, et al. J. Tissue Eng. Regen. Med. 8(11), 841-9, (2014)

Loading...


Development of a sensitive and selective liquid chromatography-mass spectrometry method for high throughput analysis of paralytic shellfish toxins using graphitised carbon solid phase extraction. Boundy MJ, Selwood AI, Harwood DT, et al. J. Chromatogr. A. 1387, 1-12, (2015)

Loading...


Chymase inhibitor-sensitive synthesis of endothelin-1 (1-31) by recombinant mouse mast cell protease 4 and human chymase. Semaan W, Desbiens L, Houde M, et al. Biochem. Pharmacol. 94(2), 91-100, (2015)

Loading...


Biomimetic collagen I and IV double layer Langmuir-Schaefer films as microenvironment for human pluripotent stem cell derived retinal pigment epithelial cells. Sorkio AE, Vuorimaa-Laukkanen EP, Hakola HM, et al. Biomaterials 51, 257-69, (2015)

Loading...


Molecularly imprinted solid-phase extraction for selective extraction of bisphenol analogues in beverages and canned food. Yang Y, Yu J, Yin J, et al. J. Agric. Food Chem. 62(46), 11130-7, (2014)

Loading...


Integrated proteomic and miRNA transcriptional analysis reveals the hepatotoxicity mechanism of PFNA exposure in mice. Wang J, Yan S, Zhang W, et al. J. Proteome Res. 14(1), 330-41, (2015)

Loading...


Evaluation of mobile phase characteristics on three zwitterionic columns in hydrophilic interaction liquid chromatography mode for liquid chromatography-high resolution mass spectrometry based untargeted metabolite profiling of Leishmania parasites. Zhang R, Watson DG, Wang L, et al. J. Chromatogr. A. 1362, 168-79, (2014)

Loading...


Bioluminescent liquid light guide pad biosensor for indoor air toxicity monitoring. Eltzov E, Cohen A, Marks RS, et al. Anal. Chem. 87(7), 3655-61, (2015)

Loading...


Comparison of capillary electrophoresis-mass spectrometry and hydrophilic interaction chromatography-mass spectrometry for anionic metabolic profiling of urine. Kok MG, Somsen GW, de Jong GJ, et al. Talanta 132, 1-7, (2014)

Loading...


Spermine oxidase maintains basal skeletal muscle gene expression and fiber size and is strongly repressed by conditions that cause skeletal muscle atrophy. Bongers KS, Fox DK, Kunkel SD, et al. Am. J. Physiol. Endocrinol. Metab. 308(2), E144-58, (2015)

Loading...


The use of doubly labeled milk protein to measure postprandial muscle protein synthesis rates in vivo in humans. Burd NA, Cermak NM, Kouw IW, et al. J. Appl. Physiol. 117(11), 1363-70, (2014)

Loading...


Bisphenol A and nonylphenol in foodstuffs: Chinese dietary exposure from the 2007 total diet study and infant health risk from formulas. Niu Y, Zhang J, Duan H, et al. Food Chem. 167, 320-5, (2014)

Loading...


Normalization of urinary pteridines by urine specific gravity for early cancer detection. Burton C, Shi H, Ma Y, et al. Clin. Chim. Acta 435, 42-7, (2014)

Loading...


Impact of approach used to determine removal levels of drugs of abuse during wastewater treatment. Rodayan A, Majewsky M, Yargeau V, et al. Sci. Total Environ. 487, 731-9, (2014)

Loading...


Sensitive and accurate liquid chromatography-tandem mass spectrometry methods for quantitative determination of a novel hepatitis C NS5B inhibitor BMS-791325 and its active metabolite in human plasma and urine. Jiang H, Demers R, Kandoussi H, et al. J. Pharm. Biomed. Anal. 107, 17-23, (2015)

Loading...


Urine, peritoneal fluid and omental fat proteomes of reproductive age women: Endometriosis-related changes and associations with endocrine disrupting chemicals. Williams KE, Miroshnychenko O, Johansen EB, et al. J. Proteomics 113, 194-205, (2014)

Loading...


Diversity of pyrrolizidine alkaloids in native and invasive Senecio pterophorus (Asteraceae): implications for toxicity. Castells E, Mulder PP, and Pérez-Trujillo M Phytochemistry 108, 137-46, (2014)

Loading...


Reversal of autism-like behaviors and metabolism in adult mice with single-dose antipurinergic therapy. Naviaux JC, Schuchbauer MA, Li K, et al. Transl. Psychiatry 4, e400, (2014)

Loading...


Synthesis of Doxorubicin loaded magnetic chitosan nanoparticles for pH responsive targeted drug delivery. Unsoy G, Khodadust R, Yalcin S, et al. Eur. J. Pharm. Sci. 62, 243-50, (2014)

Loading...


Sample preparation and UHPLC-FD analysis of pteridines in human urine. Tomšíková H, Solich P, Nováková L, et al. J. Pharm. Biomed. Anal. 95, 265-72, (2014)

Loading...


A new strategy for surface modification of polysulfone membrane by in situ imprinted sol-gel method for the selective separation and screening of L-Tyrosine as a lung cancer biomarker. Moein MM, Javanbakht M, Karimi M, et al. Analyst 140(6), 1939-46, (2015)

Loading...


Aptamer-conjugated magnetic nanoparticles for extraction of adenosine from urine followed by electrospray ion mobility spectrometry. Najafabadi ME, Khayamian T, Hashemian Z, et al. J. Pharm. Biomed. Anal. 107, 244-50, (2015)

Loading...


Adsorption-induced changes in ribonuclease A structure and enzymatic activity on solid surfaces. Wei Y, Thyparambil AA, Wu Y, et al. Langmuir 30(49), 14849-58, (2014)

Loading...


A fibrin antibody binding to fibronectin induces potent inhibition of angiogenesis. El-Ayoubi F, Amiral J, Pascaud J, et al. Thromb. Haemost. 113(1), 143-53, (2015)

Loading...


Merck 14,494

Fieser 5,15

Aldrich MSDS 1, 121:C / Corp MSDS 1 (1), 239:D / RegBook 1 (3), 3365:D / Sax 6, 263

Related Products

related product

Product #

Description

Add to Cart

Z568724 QianCap safety cap system for Ace pressure tubes and bottles, QianCap-15, fits no. 15 internal thread

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?