Synonym: Glacial acetic acid
CAS Number 64-19-7
Linear Formula CH3CO2H
Molecular Weight 60.05
Beilstein Registry Number 506007
EC Number 200-580-7
MDL number MFCD00036152
PubChem Substance ID 24860577
Popular Documents: Specification Sheet (PDF)
| Related Categories | Acetic Acid, Acids & Bases, Essential Chemicals, Research Essentials |
| vapor density | 2.07 (vs air) |
| assay | ≥99.99% (Purity based on metals analysis) |
| autoignition temp. | 800 °F |
| expl. lim. | 16 %, 92 °F |
| 4 %, 59 °F | |
| refractive index |
n |
| bp | 117-118 °C(lit.) |
25, 100, 500 mL in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?