EMAIL THIS PAGE TO A FRIEND

33936 Sigma-Aldrich

Hydroxynaphthol blue

indicator for metal titration

Synonym: 1-(2-Hydroxy-4-sulfo-1-naphthylazo)-2-naphthol-3,6-disulfonic acid trisodium salt, 3-Hydroxy-4-(2-hydroxy-4-sulfo-1-naphthylazo)naphthalene-2,7-disulfonic acid trisodium salt

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Indicators, Metal Titration Indicators, Titration More...
grade   indicator for metal titration
suitability   complies for suitability for complexometry

Description

General description

Visit our Titration Center to learn more.

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




Explore the New Analytical Standards Portal

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Colorimetric detection of loop-mediated isothermal amplification reaction by using hydroxy naphthol blue. Goto M, Honda E, Ogura A, et al. Biotechniques 46(3), 167-72, (2009)

Loading...


Rapid detection of Pseudomonas aeruginosa in mouse feces by colorimetric loop-mediated isothermal amplification. Goto M, Shimada K, Sato A, et al. J. Microbiol. Methods 81(3), 247-52, (2010)

Loading...


[Colorimetric detection of human influenza A H1N1 virus by reverse transcription loop mediated isothermal amplification]. Nie K, Wang DY, Qin M, et al. Bing Du Xue Bao 26(2), 81-7, (2010)

Loading...


Development of a loop-mediated isothermal amplification assay for rapid detection of capripoxviruses. Das A, Babiuk S, and McIntosh MT J. Clin. Microbiol. 50(5), 1613-20, (2012)

Loading...


Rapid detection of Opisthorchis viverrini copro-DNA using loop-mediated isothermal amplification (LAMP). Arimatsu Y, Kaewkes S, Laha T, et al. Parasitol. Int. 61(1), 178-82, (2012)

Loading...


Visual detection of pandemic influenza A H1N1 Virus 2009 by reverse-transcription loop-mediated isothermal amplification with hydroxynaphthol blue dye. Ma XJ, Shu YL, Nie K, et al. J. Virol. Methods 167(2), 214-7, (2010)

Loading...


Visual detection of high-risk human papillomavirus genotypes 16, 18, 45, 52, and 58 by loop-mediated isothermal amplification with hydroxynaphthol blue dye. Luo L, Nie K, Yang MJ, et al. J. Clin. Microbiol. 49(10), 3545-50, (2011)

Loading...


Visual detection of turkey coronavirus RNA in tissues and feces by reverse-transcription loop-mediated isothermal amplification (RT-LAMP) with hydroxynaphthol blue dye. Cardoso TC, Ferrari HF, Bregano LC, et al. Mol. Cell Probes 24(6), 415-7, (2010)

Loading...


Visual detection of human enterovirus 71 subgenotype C4 and Coxsackievirus A16 by reverse transcription loop-mediated isothermal amplification with the hydroxynaphthol blue dye. Nie K, Zhang Y, Luo L, et al. J. Virol. Methods 175(2), 283-6, (2011)

Loading...


Visual detection of the human metapneumovirus using reverse transcription loop-mediated isothermal amplification with hydroxynaphthol blue dye. Wang X, Zhang Q, Zhang F, et al. Virol. J. 9, 138, (2012)

Loading...


Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?