EMAIL THIS PAGE TO A FRIEND
33971 Sigma-Aldrich

1,2-Dibromobenzene

purum, ≥97.0% (GC)

  • CAS Number 583-53-9

  • Empirical Formula (Hill Notation) C6H4Br2

  • Molecular Weight 235.90

  •  Beilstein Registry Number 970241

  •  EC Number 209-507-3

  •  MDL number MFCD00000057

  •  PubChem Substance ID 329754712

Purchase

Properties

Related Categories Aryl, Building Blocks, C6, Chemical Synthesis, Halogenated Hydrocarbons,
vapor density   8.2 (vs air)
InChI Key   WQONPSCCEXUXTQ-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
color   brown-yellow
refractive index   n20/D 1.611(lit.)
  n20/D 1.611
bp   224 °C(lit.)
mp   4-6 °C(lit.)
density   1.956 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
197.6 °F
Flash Point(C) 
92 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

D39002 1,2-Dibromobenzene, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?