EMAIL THIS PAGE TO A FRIEND
33990 Sigma-Aldrich

1,4-Dibromobenzene

purum, ≥97.0% (GC)

  • CAS Number 106-37-6

  • Empirical Formula (Hill Notation) C6H4Br2

  • Molecular Weight 235.90

  •  Beilstein Registry Number 1904543

  •  EC Number 203-390-2

  •  MDL number MFCD00000089

  •  PubChem Substance ID 329754729

Purchase

Properties

Related Categories Aryl, Building Blocks, C6, Chemical Synthesis, Halogenated Hydrocarbons,
InChI Key   SWJPEBQEEAHIGZ-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
bp   219 °C(lit.)
mp   83-87 °C(lit.)
  85-89 °C
density   1.841 g/mL at 25 °C(lit.)
Gene Information   human ... CYP2A6(1548)
mouse ... Cyp2a5(13087)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
CZ1791000
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

D39029 1,4-Dibromobenzene, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?