• USA Home
  • 33995 - 4,4′-Dibromobenzophenone

EMAIL THIS PAGE TO A FRIEND
33995 Sigma-Aldrich

4,4′-Dibromobenzophenone

purum, ≥98.0% (GC)

  • CAS Number 3988-03-2

  • Linear Formula (BrC6H4)2CO

  • Molecular Weight 340.01

  •  Beilstein Registry Number 2049958

  •  EC Number 223-632-0

  •  MDL number MFCD00016330

Purchase

Properties

Related Categories Building Blocks, C13 to C14, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   LFABNOYDEODDFX-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
mp   171-174 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

166634 4,4′-Dibromobenzophenone, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?