CAS Number 7664-41-7
Linear Formula NH3
Molecular Weight 17.03
MDL number MFCD00011418
PubChem Substance ID 24860942
Popular Documents: Specification Sheet (PDF)
| concentration | 2.0 M in methanol |
| density | 0.787 g/mL at 25 °C |
100, 800 mL in Sure/Seal™
20 L in steel drum
2 L in glass bottle
5 mL in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?