• USA Home
  • 34187 - Dibenzyl phthalate-3,4,5,6-d4

EMAIL THIS PAGE TO A FRIEND
34187 Sigma-Aldrich

Dibenzyl phthalate-3,4,5,6-d4

analytical standard

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Aromatics, Chemical Class,
grade   analytical standard
InChI Key   UCVPKAZCQPRWAY-ZZRPVTOQSA-N
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
mass shift   M+4

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?