EMAIL THIS PAGE TO A FRIEND
34215 Sigma-Aldrich

Dibromomaleic acid

purum, ≥97.0% (T)

  • CAS Number 608-37-7

  • Linear Formula HO2CC(Br)=C(Br)CO2H

  • Molecular Weight 273.86

  •  Beilstein Registry Number 1724799

  •  EC Number 210-162-6

  •  MDL number MFCD00004207

  •  PubChem Substance ID 24861032

Purchase

Properties

Related Categories Building Blocks, C1 to C5, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   PMNMPRXRQYSFRP-UPHRSURJSA-N
grade   purum
assay   ≥97.0% (T)
mp   ~125 °C (dec.)(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?