EMAIL THIS PAGE TO A FRIEND
342661 Sigma-Aldrich

Aluminum oxide

primarily α-phase, fused, 200-325 mesh, ≥99%

Synonym: Alumina

Purchase

Properties

InChI Key   TWNQGVIAIRXVLR-UHFFFAOYSA-N
assay   ≥99%
form   fused
particle size   200-325 mesh
mp   2040 °C(lit.)

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
BD1200000
Protocols & Articles

Articles

Advanced Inorganic Materials for Solid State Lighting

In 1907, the first electroluminescent device, with silicon carbide (Aldrich Prod. Nos. 594911, 378097, 357391) as the active material, was described by Henry Round as displaying a “bright glow”.1,2 L...
Alexander Birkel, Kristin A. Denault, Nathan C. George, Ram Seshadri
Material Matters, Volume 7, Number 2, 22–27
Keywords: Capillary electrophoresis, Chromatin immunoprecipitation, Combustion, Ligands, Microwave synthesis, Sol-gel, Substitutions

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?