CAS Number 1309-48-4
Linear Formula MgO
Molecular Weight 40.30
EC Number 215-171-9
MDL number MFCD00011109
PubChem Substance ID 24860734
| Related Categories | Catalysis and Inorganic Chemistry, Chemical Synthesis, Magnesium, Materials Science, Metal and Ceramic Science, |
| assay | 99.9% trace metals basis |
| form | chips |
| fused | |
| particle size | -4 mesh |
| mp | 2852 °C(lit.) |
100 g in glass bottle
Suitable for vacuum deposition.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?