Synonym: Ferric oxide
CAS Number 1309-37-1
Empirical Formula (Hill Notation) Fe2O3
Molecular Weight 159.69
EC Number 215-168-2
MDL number MFCD00011008
PubChem Substance ID 24861096
Popular Documents: Specification Sheet (PDF)
| Related Categories | Iron, Materials Science, Metal and Ceramic Science, Oxides |
| assay | 99.8% trace metals basis |
| form | pieces |
| sintered | |
| particle size | 3-12 mm |
50 g in poly bottle
Suitable for vacuum deposition.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?