• USA Home
  • 34305 - Ethyl 2,3-dibromopropionate

EMAIL THIS PAGE TO A FRIEND
34305 Sigma-Aldrich

Ethyl 2,3-dibromopropionate

purum, ≥95.0% (GC)

Purchase

Properties

Related Categories Building Blocks, C2 to C5, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   OENICUBCLXKLJQ-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
refractive index   n20/D 1.5(lit.)
bp   211-214 °C/746 mmHg(lit.)
density   1.788 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2922 6.1(8) / PGII
WGK Germany 
2
RTECS 
UA2458382
Flash Point(F) 
195.8 °F
Flash Point(C) 
91 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

E22805 Ethyl 2,3-dibromopropionate, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?