EMAIL THIS PAGE TO A FRIEND
34319 Sigma-Aldrich

Chlorfenethol

PESTANAL®, analytical standard

Synonym: 1,1-Bis(4-chlorophenyl)ethanol

  • CAS Number 80-06-8

  • Linear Formula (ClC6H4)2C(CH3)OH

  • Molecular Weight 267.15

  •  Beilstein Registry Number 2505345

  •  EC Number 201-246-3

  •  MDL number MFCD00000628

  •  PubChem Substance ID 329754943

Purchase

Properties

Related Categories Acaricides, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, C,
grade   analytical standard
InChI Key   URYAFVKLYSEINW-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   69-71 °C(lit.)
format   neat
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
DC7875000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?