EMAIL THIS PAGE TO A FRIEND
34337 Sigma-Aldrich

Dinoterb acetate

PESTANAL®, analytical standard

  • CAS Number 3204-27-1

  • Empirical Formula (Hill Notation) C12H14N2O6

  • Molecular Weight 282.25

  •  Beilstein Registry Number 2163325

  •  EC Number 221-706-7

  •  MDL number MFCD00129667

  •  PubChem Substance ID 329754951

Purchase

Properties

Related Categories Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, D, Dinitrophenol,
grade   analytical standard
InChI Key   BMJYKXALMDAIEG-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Caution

Restricted to professional users. Attention − Avoid exposure − obtain special instructions before use.

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
SK0175000
Flash Point(F) 
>212 °F
Flash Point(C) 
>100 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?