EMAIL THIS PAGE TO A FRIEND
34354 Sigma-Aldrich

Methoxyphenone

PESTANAL®, analytical standard

Synonym: 3,3′-Dimethyl-4-methoxybenzophenone

  • CAS Number 41295-28-7

  • Empirical Formula (Hill Notation) C16H16O2

  • Molecular Weight 240.30

  •  Beilstein Registry Number 1966899

  •  EC Number 255-300-6

  •  MDL number MFCD00128925

  •  PubChem Substance ID 329754962

Purchase

Properties

Related Categories Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, H-M, Pesticides & Metabolites Standards More...
grade   PESTANAL®, analytical standard
InChI Key   BWPYBAJTDILQPY-UHFFFAOYSA-N
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
PC4961000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?