• USA Home
  • 34374 - 1,2,3,4-Tetrachloro-5-nitrobenzene

EMAIL THIS PAGE TO A FRIEND
34374 Sigma-Aldrich

1,2,3,4-Tetrachloro-5-nitrobenzene

PESTANAL®, analytical standard

  • CAS Number 879-39-0

  • Empirical Formula (Hill Notation) C6HCl4NO2

  • Molecular Weight 260.89

  •  Beilstein Registry Number 2273256

  •  EC Number 212-906-5

  •  MDL number MFCD00007064

  •  PubChem Substance ID 329754969

Purchase

Properties

Related Categories Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, Pesticides & Metabolites Standards, Q-Z More...
grade   analytical standard
InChI Key   MTBYTWZDRVOMBR-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DB9625000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?